Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SPINeasy™ DNA Purification Kit

Catalog No. MP116548050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116548050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116548050 Supplier MP Biomedicals™ Supplier No. 116548050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for fast and efficient cleanup of pre-purified DNA samples. Removes a wide range of inhibitory compounds, such as those found in environmental samples (humic acid, heavy metals, etc.), as well as organic compounds that may co-purify with DNA from other extraction procedures.

  • The SPINeasy DNA Purification Kit is designed for the removal of contaminants that may co-purify with DNA and inhibit downstream enzymatic, PCR applications.
  • Inhibitory compounds such as those found in environmental samples (humic acid, heavy metals, etc.), as well as organic compounds that may co-purify with DNA from other extraction procedures, are efficiently and quickly removed using this kit.
  • The kit offers two choices of binding buffers. Buffer P1 is recommended for purification of samples with slight contamination of humic acids (light brownish color) or other PCR inhibitors. Buffer P1HA contains a novel and proprietary humic acid removal technology, which is recommended for processing samples contaminated with a considerable amount of humic substances (strong brownish color).
  • The kit has been validated with genomic DNA isolated from various problematic soil samples, as well as with DNA samples spiked with humic acid molecules. The data shows that the kit can clean up to 50 µg DNA with a size ranging from 200 bp to high molecular weight, with expected recovery of > 80%.
  • Cleaned-up DNA samples can be readily used in downstream applications, including long fragment PCR, qPCR, and next-generation sequencing (16S and whole genome).
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Format Kit
Isolation Technology Spin Column
Sample Type 25 to 200 µL DNA preparations > 200 bp
Includes Buffer P1 50 mL, Buffer P1HA 50mL, Buffer P2 40 mL, Buffer P3 40 mL, Buffer P4 6 mL, 50x Column P, 100 x 2.0 mL collection tubes, 50 x 1.5 mL collection tubes
Product Type DNA Purification Kit
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.