Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ DNA/RNA Kit for Blood

Catalog No. MP116582050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116582050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116582050 Supplier MP Biomedicals Supplier No. 116582050
Add to Cart
Add to Cart

Provides a fast and efficient workflow for reliable recovery of both DNA and RNA from blood samples.

  • Optimized for simultaneous isolation of genomic DNA and RNA from whole blood samples, effectively addressing challenges related to inhibitors and low nucleic acid concentrations.
  • Based on high-performance silica membrane spin-column technology to ensure superior yield and purity of both DNA and RNA.
  • Supports rapid extraction from whole blood preserved with different anticoagulants through a streamlined protocol completed in a short processing time.
  • Utilizes Lysis Buffer SL1 in combination with a minimal amount of solvent to eliminate the need for excessive homogenization while efficiently separating nucleic acids from organic impurities such as hemoglobin, immunoglobulins, and other blood proteins.
  • Enables co-purification of DNA and RNA using two distinct columns from the same sample, ensuring a hassle-free, unbiased, and highly time-efficient workflow.
  • Eliminates the requirement for enzymatic treatments, including DNase I or RNase A digestion, allowing maximum recovery of both nucleic acids.
  • Employs a simplified extraction system using a single solvent and one additional binding buffer, reducing handling complexity.
  • Optimized washing steps effectively remove salts, metabolites, and macromolecular cellular components.
  • Compatible with both fresh and frozen whole blood samples and delivers high-quality DNA and RNA suitable for downstream applications such as PCR amplification.
TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
Isolation Technology Silica Spin Column
Scale Mini
Sample Type Blood
No. of Reactions 50 Preps
Product Type DNA/RNA Kit for Blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.