Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ DNA/RNA Kit for Feces

Catalog No. MP116555050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116555050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116555050 Supplier MP Biomedicals Supplier No. 116555050
Add to Cart
Add to Cart

Enables efficient recovery of high-quality microbial nucleic acids from complex fecal samples.

  • Designed for the simultaneous purification of microbiome genomic DNA and RNA from a wide range of fecal sample types, regardless of sample composition.
  • Delivers highly pure nucleic acids free from inhibitory contaminants, eliminating the need for additional inhibitor removal steps.
  • Produces DNA and RNA that are immediately suitable for downstream molecular applications, including qPCR and RT-qPCR.
TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
Isolation Technology Silica Spin Column
Scale Mini
No. of Reactions 50 Preps
Product Type DNA/RNA Kit for Feces
Species All
Target Total Nucleic Acids
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.