Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SPINeasy™ DNA/RNA/Protein All-In-One Kit

Catalog No. MP116544050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116544050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116544050 Supplier MP Biomedicals™ Supplier No. 116544050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fast and efficient isolation of DNA, RNA and protein from a single sample using spin column technology. Lysing Matrix A for thorough and complete tissue lysis with FastPrep instruments.

  • SPINeasy DNA/RNA/Protein All-In-One Kit utilizes a convenient workflow and silica membrane spin columns to isolate DNA, RNA, and protein components from the same sample, without the use of toxic substances such as phenol and chloroform.
  • The use of our specially formulated Lysis Buffer R and Lysing Matrix A in combination with FastPrep instruments from MP Biomedicals enables highly efficient lysis of tissue samples within seconds.
  • DNA, RNA, and proteins are then sequentially purified from the same lysate.
  • Briefly, DNA is absorbed onto the first spin column, while the flow-through is collected and used for RNA purification.
  • The second spin column captures RNA and the flow-through from this step is used for protein extraction.
  • DNA and RNA are bound on the first and second columns, respectively, and then washed and eluted.
  • In the final extraction, proteins are precipitated out of solution, pelleted down by centrifugation, washed, and resuspended.
  • Each molecular component is then immediately available for their respective downstream applications.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Format Kit
Isolation Technology Spin Column
Sample Type Blood, Cell Culture, Animal Tissue, Swabs
Includes 50 x Lysing Matrix A, 60 mL Lysis Buffer R, 30 mL Wash Buffer D, 12 mL Wash Buffer R, 10 mL Elution Buffer GD, 10 mL Nuclease-free water, 1 vial DNase I, 5 mL DNase I Buffer, 80 mL Protein Precipitant, 100 x Column A with collection tube
Product Type Isolation Kit
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.