Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SPINeasy™ Plasmid Miniprep Kit

Catalog No. MP116534050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116534050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116534050 Supplier MP Biomedicals™ Supplier No. 116534050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spin column technology for fast, high-quality plasmid DNA isolation from bacterial cultures. High purity plasmid DNA, free of gDNA contamination following a modified alkaline lysis method. Plasmid DNA suitable for a variety of downstream applications, including cloning, restriction digestion, and transformation.

  • SPINeasy Plasmid Miniprep Kit is a silica membrane spin column kit that enables the purification of up to 20 µg of plasmid DNA from 1 to 5 mL of transformed bacterial cultures.
  • High-purity plasmid DNA can be obtained within 25 minutes through a simple purification process, immediately ready for routine molecular biology applications including cloning, restriction digestion, PCR, and in-vitro transcription.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Format Kit
Isolation Technology Spin Column
Includes 20 mL Resuspension Buffer SN1, 20 mL Alkaline Lysis Buffer SN2, 30 mL Neutralization Buffer SN3, 21 mL Wash Buffer NW1, 6 mL Wash Buffer NW2, 10 mL Elution Buffer SN, 200 µL RNase A, 50 x Column SN with collection tubes
Product Type Plasmid DNA Purification Kit
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.