Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ RNA Kit for Soil

Catalog No. MP116585050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116585050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116585050 Supplier MP Biomedicals Supplier No. 116585050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Utilizes silica-membrane spin-column technology for efficient and high-purity RNA extraction.

  • Works with a variety of soil types, including those with high humic acid content and heavy contaminants.
  • Provides rapid extraction in under 1 hour without the use of hazardous chemicals.
  • Delivers high-quality RNA suitable for sensitive applications like reverse transcription, real-time PCR, and sequencing.
TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer
Control Type RNA Control
Isolation Technology Silica Spin Column
Species All
Sample Type Soil
No. of Reactions 50 Preps
Product Type RNA kit for Soil
Target Total RNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.