Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ Yeast Plasmid Miniprep Kit

Catalog No. MP116584050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116584050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116584050 Supplier MP Biomedicals Supplier No. 116584050
Add to Cart
Add to Cart

Utilizes silica-membrane spin-column technology for high-performance yeast plasmid extraction.

  • High-performance plasmid extraction kit using silica membrane spin column technology.
  • Processes up to 1.5 mL cultures to maximize plasmid yield.
  • Combines enzymatic and chemical lysis methods to streamline the workflow and reduce total handling time to under 50 minutes.
TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
Isolation Technology Silica Spin Column
Scale Mini
No. of Reactions 50 Preps
Product Type Yeast Plasmid Miniprep Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.