Learn More
Aviva Systems Biology Corp SPINK1 Antibody - N-terminal region (ARP33865_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33865_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPINK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SPINK1. Alternate names: PCTT, PSTI, Spink3, TATI, TCPApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-014-55180