Learn More
Sino Biological SPINK4 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11669R265100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SPINK4 (rh SPINK4; Catalog#11669-H08H; NP_055286.1; Met1-Cys86).
Catalog No. 50-224-7787