Learn More
Chemglass Life Sciences Spinner Flask, 100mL, Jacketed, BioProcess, Plain Bottom, Complete
SDP
Catalog No. 5000980596
Encompass_Preferred
Water jacketed, flat bottomed spinner flasks consist of a water jacketed spinner flask, manufactured from Type 1 Class A borosilicate glass. Flasks are typically used with a circulating water bath. Jacket volumes are approximate and flasks are not graduated. Note: Paddle and Cap assembly are not included.
Catalog No. 50-009-80596 Supplier Chemglass Life Sciences Supplier No. CLS1430J100
Water jacketed, flat bottomed spinner flasks consist of a water jacketed spinner flask, manufactured from Type 1 Class A borosilicate glass. Flasks are typically used with a circulating water bath. Jacket volumes are approximate and flasks are not graduated. Note: Paddle and Cap assembly are not included.