Learn More
Chemglass Life Sciences Spinner Flask, 3000mL, Jacketed, BioProcess, Plain Bottom, Complete
SDP
Catalog No. 5000083162
Encompass_Preferred
Water jacketed, flat bottomed spinner flasks consist of a water jacketed spinner flask, manufactured from Type 1 Class A borosilicate glass.Flasks are typically used with a circulating water bath.Jacket volumes are approximate and flasks are not graduated.Note: Paddle and Cap assembly are not included.
Catalog No. 50-000-83162 Supplier Chemglass Life Sciences Supplier No. CLS1430J3L
Water jacketed, flat bottomed spinner flasks consist of a water jacketed spinner flask, manufactured from Type 1 Class A borosilicate glass.Flasks are typically used with a circulating water bath.Jacket volumes are approximate and flasks are not graduated.Note: Paddle and Cap assembly are not included.