Learn More
Alkali Scientific Spinner Flask Flat Bottom, 100mL
SDP
Catalog No. 5000662334
Encompass_Preferred
Flat bottom spinner flasks promote optimal growth of suspension cultures. The complete system includes a flask with two angled sidearms and a PTFE paddle with glass shaft that adjusts through the compression fitting in the cap. Flat bottom vessel 100mL, 250mL, 500mL and 1L. Heavy wall center port and side arms. FDA approved materials. Capacity (mL) Center Neck Sidearm OD x OAH (mm) 100 70 GL 32 70 x 175 250 70 GL 32 85 x 210 500 70 GL 45 110 x 250 1000 100 GL 45 135 x 305 3000 100 GL 45 165 x 355 Spinner flasks are constructed of high quality components with the following specifications: Vessels: Borosilicate type 1 glass Shafts: Borosilicate glass 7mm Magnets: PTFE encapsulated Paddle: PTFE Cap: Polypropylene Compression fitting: Polypropylene Liner: Silicone O ring: Silicone
Catalog No. 50-006-62334 Supplier Alkali Scientific Supplier No. CTS100
Flat bottom spinner flasks promote optimal growth of suspension cultures. The complete system includes a flask with two angled sidearms and a PTFE paddle with glass shaft that adjusts through the compression fitting in the cap. Flat bottom vessel 100mL, 250mL, 500mL and 1L. Heavy wall center port and side arms. FDA approved materials. Capacity (mL) Center Neck Sidearm OD x OAH (mm) 100 70 GL 32 70 x 175 250 70 GL 32 85 x 210 500 70 GL 45 110 x 250 1000 100 GL 45 135 x 305 3000 100 GL 45 165 x 355 Spinner flasks are constructed of high quality components with the following specifications: Vessels: Borosilicate type 1 glass Shafts: Borosilicate glass 7mm Magnets: PTFE encapsulated Paddle: PTFE Cap: Polypropylene Compression fitting: Polypropylene Liner: Silicone O ring: Silicone