Learn More
Medchemexpress LLC Spinorphin (TFA) | 2828433-21-0 | 99.4% | 991.06 | 5 MG
SDP
Catalog No. 5000423661
Encompass_Preferred
Spinorphin TFA is an inhibitor of enkephalin-degrading enzymes. It inhibits aminopeptidase, dipeptidyl aminopeptidase III, angiotensin-converting enzyme, and enkephalinase. Spinorphin also has an antinociceptive effect.
  • For research use only
Catalog No. 50-004-23661 Supplier Medchemexpress LLC Supplier No. HYP1044A5MG
Spinorphin TFA is an inhibitor of enkephalin-degrading enzymes. It inhibits aminopeptidase, dipeptidyl aminopeptidase III, angiotensin-converting enzyme, and enkephalinase. Spinorphin also has an antinociceptive effect.
  • For research use only