Learn More
Chem Service Inc Spinosad,168316-95-8,100MG
SDP
Catalog No. NC9860526
Encompass_Preferred
Molecular Formula C42H67NO10, Purity 96.80%, Molecular Weight 746, Melting Point 161.5 - 170 °C
Catalog No. NC9860526 Supplier Chem Service Inc Supplier No. N13222100MG
Molecular Formula C42H67NO10, Purity 96.80%, Molecular Weight 746, Melting Point 161.5 - 170 °C