Learn More
Medchemexpress LLC Spinosyn D | 131929-63-0 | 99.0% | 746.0 g/mol | C42H67NO10 | 1MG
SDP
Catalog No. 5000235981
Encompass_Preferred
Spinosyn D is a naturally occurring spinosyn-class insecticide and a major component of spinosad. Supplied as a high-purity research reagent, it is used as a reference standard and for studies of insecticidal activity and mode of action.
Catalog No. 50-002-35981 Supplier Medchemexpress LLC Supplier No. HY1253261MG
Spinosyn D is a naturally occurring spinosyn-class insecticide and a major component of spinosad. Supplied as a high-purity research reagent, it is used as a reference standard and for studies of insecticidal activity and mode of action.