Learn More
Cayman Chemical Spinosyn D 17-pseudoaglycone, Purity: ≥95%, CAS: 131929-55-0, 2.5 mg
SDP
Catalog No. 502939447
Encompass_Preferred
A hydrolysis product of spinosyn D; not lethal to tobacco budworms (H. virescens) at concentrations up to 64 ppm
Catalog No. 50-293-9447 Supplier Cayman Chemical Supplier No. 279752.5 MG
A hydrolysis product of spinosyn D; not lethal to tobacco budworms (H. virescens) at concentrations up to 64 ppm