Learn More
Cayman Chemical SpInosyn D aglycon 2.5mg
SDP
Catalog No. 502940899
Encompass_Preferred
An aglycone form of spinosyn D; not lethal to H. virescens (tobacco budworm) neonate larvae (LC50 = >64 ppm)
Catalog No. 50-294-0899 Supplier Cayman Chemical Supplier No. 279372.5 MG
An aglycone form of spinosyn D; not lethal to H. virescens (tobacco budworm) neonate larvae (LC50 = >64 ppm)