Learn More
Aviva Systems Biology Corp SPINT1 Antibody - C-terminal region (ARP59226_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59226_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPINT1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human SPINT1. Alternate names: HAI, HAI1, MANSC2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-622