Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SPINT1 (HAI-1) ELISA Kit

Catalog No. EHSPINT1X5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHSPINT1X5 5 x 96 Tests
EHSPINT1 96 Tests
EHSPINT1X10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHSPINT1X5 Supplier Invitrogen™ Supplier No. EHSPINT1X5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human SPINT1 (HAI-1) ELISA quantitates Hu SPINT1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu SPINT1. Principle of the method The Human SPINT1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43278
Assay Range 0.819-200 ng/mL
Assay Sensitivity 0.8 ng/mL
Calibration Range 0.8 ng/mL to 200 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cell Structure/Organelle Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human HAI-1, no cross-reactivity with mouse HAI-1, human APP, GASP-1, GASP-2, HAI-2 A, TFPI, TFPI-2 is observed.
Sufficient For 5 x 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias HAI; HAI1; HAI-1; hepatocyte growth factor activator inhibitor 1; hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; kunitz-type protease inhibitor 1; MANSC2; serine peptidase inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 1; Spint1; UNQ223/PRO256HAI; HAI1; HAI-1; hepatocyte growth factor activator inhibitor 1; hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; kunitz-type protease inhibitor 1; MANSC2; serine peptidase inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 1; Spint1; UNQ223/PRO256; HAI; HAI1; MANSC2
Gene ID (Entrez) 6692
Gene Symbol SPINT1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Hepatocyte growth factor activator inhibitor type 1, HAI-1
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Extracellular Matrix & Cell Adhesion
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym UNQ223/PRO256, HAI, HAI1, MANSC2
Target HAI-1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.