Learn More
Aviva Systems Biology Corp SPINT2 Antibody - middle region (ARP49615_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49615_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPINT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SPINT2. Alternate names: HAI-2, HAI2, Kop, PB, DIAR3ApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-014-60124