Learn More
Aviva Systems Biology Corp SPINT2 Antibody - middle region (ARP59164_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59164_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPINT2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SPINT2. Alternate names: DIAR3, FLJ45571, HAI-2, HAI2, Kop, PBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-602