Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Spinvane
GREENER_CHOICESDP

Catalog No. 31502134 Shop All Kemtech Products
Change view
Click to view available options
Shape:
Triangle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shape
31-502-134 Triangle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-502-134 Supplier Synthware™ Supplier No. S470001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Magnetic Spinvane for conical shaped mini-vials.

  • Magnet is encased in PTFE and is used with conical shaped Mini-Vials.

Specifications

Length (Metric) 305 mm
For Use With (Equipment) 3.0 and 5.0 Conical Bottom Mini Vials
Coating PTFE
Autoclavable Autoclavable
Type Magnetic Stir Bar
Shape Triangle
Quantity 1 Ea.
For Use With (Application) Organic Synthesis
Color White
Material PTFE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.