Learn More
V&P Scientific Inc SpinVessel, 50mL, 25/pack or 500 pack
Catalog No. NC2125714
Encompass
SpinVessel, 50mL, Conical Bottom, Disposable, Sterile, RNase, DNase and DNA Free, Nonpyrongenic Virgin Polypropylene, with Pulsed Radial Flow Spin Fins and Projections, US Patent #11,623,188, European Patent #3887049
This item is not returnable. View return policy
Catalog No. NC2125714 Supplier V&P Scientific Inc Supplier No. VP 830SV50PK
SpinVessel, 50mL, Conical Bottom, Disposable, Sterile, RNase, DNase and DNA Free, Nonpyrongenic Virgin Polypropylene, with Pulsed Radial Flow Spin Fins and Projections, US Patent #11,623,188, European Patent #3887049