Learn More
V&P Scientific Inc SpinVessel, 50mL, 25/pack or 500 pack
Supplier:  V&P Scientific Inc VP 830SV50
Encompass
SpinVessel, 50mL, Conical Bottom, Disposable, Sterile, RNase, DNase and DNA Free, Nonpyrongenic Virgin Polypropylene, with Pulsed Radial Flow Spin Fins and Projections, US Patent #11,623,188, European Patent #3887049
Catalog No. NC2125718