Learn More
Medchemexpress LLC Spiperone hydrochloride | 2022-29-9 | 99.0% | 5 MG
SDP
Catalog No. 5000413662
Encompass_Preferred
Spiperone hydrochloride (Spiroperidol hydrochloride) is a selective antagonist for the dopamine D2 receptor (with Ki values of 0.06 nM, 0.6 nM, 0.08 nM, ~350 nM, and ~3500 nM for D2, D3, D4, D1, and D5 receptors, respectively) and the 5-HT2A/5-HT1A receptors (with Kis of 1 nM/49 nM). It also acts as a selective α1B-adrenoceptor antagonist and activates the calcium-activated chloride channel (CaCC). This compound exhibits antipsychotic and anti-inflammatory activities and is intended for research use only.
  • Selective dopamine D2 receptor antagonist
  • Antagonizes 5-HT2A/5-HT1A receptors
  • Selective α1B-adrenoceptor antagonist
  • Activates calcium-activated chloride channel (CaCC)
  • Demonstrates antipsychotic activity
  • Exhibits anti-inflammatory activities
Catalog No. 50-004-13662 Supplier Medchemexpress LLC Supplier No. HYB1371A5MG
Spiperone hydrochloride (Spiroperidol hydrochloride) is a selective antagonist for the dopamine D2 receptor (with Ki values of 0.06 nM, 0.6 nM, 0.08 nM, ~350 nM, and ~3500 nM for D2, D3, D4, D1, and D5 receptors, respectively) and the 5-HT2A/5-HT1A receptors (with Kis of 1 nM/49 nM). It also acts as a selective α1B-adrenoceptor antagonist and activates the calcium-activated chloride channel (CaCC). This compound exhibits antipsychotic and anti-inflammatory activities and is intended for research use only.
  • Selective dopamine D2 receptor antagonist
  • Antagonizes 5-HT2A/5-HT1A receptors
  • Selective α1B-adrenoceptor antagonist
  • Activates calcium-activated chloride channel (CaCC)
  • Demonstrates antipsychotic activity
  • Exhibits anti-inflammatory activities