Learn More
Grainger Chicago-Latrobe Spiral Flute Tap, 3/8"-24 UNF, H3/2B, High Speed Steel, 1 ea
Catalog No. 502755004
Encompass_Preferred
This high speed steel spiral flute tap is designed for efficient threading in various materials including steel, aluminum, brass, bronze, and copper. Featuring a 3/8 in.-24 UNF thread size, this tool includes an extended neck and a bottoming chamfer to ensure precise results in blind hole applications.
  • High speed steel construction for durability
  • Spiral flute design with 52° helix angle
  • Bright uncoated finish
  • Right hand thread direction
  • H3 pitch diameter limit and 2B class of fit
  • 3 flutes for effective chip evacuation
  • Overall length of 2 15/16 in. with a 3/4 in. thread length
  • Manufactured to USCTI Table 302A standards
Catalog No. 50-275-5004 Supplier Grainger Supplier No. 15J967
This high speed steel spiral flute tap is designed for efficient threading in various materials including steel, aluminum, brass, bronze, and copper. Featuring a 3/8 in.-24 UNF thread size, this tool includes an extended neck and a bottoming chamfer to ensure precise results in blind hole applications.
  • High speed steel construction for durability
  • Spiral flute design with 52° helix angle
  • Bright uncoated finish
  • Right hand thread direction
  • H3 pitch diameter limit and 2B class of fit
  • 3 flutes for effective chip evacuation
  • Overall length of 2 15/16 in. with a 3/4 in. thread length
  • Manufactured to USCTI Table 302A standards