Learn More
Grainger Balax Spiral Flute Tap, Powdered Metal, 2-56 STI, 2 Flutes, 1 ea
Catalog No. 502785026
Encompass_Preferred
This spiral-flute machine tap features an open spiral design to efficiently eject chips upward, preventing packing within blind holes. It is designed for installation into toolholders on CNC or similar machinery for high-performance threading applications.
  • Constructed from powdered metal with a bright finish
  • Right hand thread direction with a 45.00° helix angle
  • Suitable for machining bronze, aluminum, stainless steel, brass, carbon steel, magnesium, and copper
  • Features 2 flutes and a modified bottoming chamfer
  • Thread size: #2-56 UNC with H2 thread limit and 2B class of fit
  • Overall length: 1-13/16 in., thread length: 0.500 in., and shank diameter: 0.141 in.
Catalog No. 50-278-5026 Supplier Grainger Supplier No. 196YP6
This spiral-flute machine tap features an open spiral design to efficiently eject chips upward, preventing packing within blind holes. It is designed for installation into toolholders on CNC or similar machinery for high-performance threading applications.
  • Constructed from powdered metal with a bright finish
  • Right hand thread direction with a 45.00° helix angle
  • Suitable for machining bronze, aluminum, stainless steel, brass, carbon steel, magnesium, and copper
  • Features 2 flutes and a modified bottoming chamfer
  • Thread size: #2-56 UNC with H2 thread limit and 2B class of fit
  • Overall length: 1-13/16 in., thread length: 0.500 in., and shank diameter: 0.141 in.