Learn More
Grainger WIDIA Spiral Flute Tap, #5-40 UNC, H2, 3B, TiCN
Catalog No. 502772469
Encompass_Preferred
This spiral flute tap is designed for efficient threading in blind hole applications. Constructed from cobalt high-speed steel and finished with a TiCN coating, it provides increased durability and abrasion resistance for diverse material compatibility.
  • Designed for blind hole tapping
  • Suitable for steel, stainless steel, aluminum, brass, bronze, copper, and cast iron
  • Features a 45° helix angle for effective chip evacuation
  • Coated with TiCN for enhanced performance
  • Includes an extended neck design
  • ANSI standard dimensions with H2 pitch diameter limits and 3B class of fit
  • Straight shank with square drive
Catalog No. 50-277-2469 Supplier Grainger Supplier No. 13X404
This spiral flute tap is designed for efficient threading in blind hole applications. Constructed from cobalt high-speed steel and finished with a TiCN coating, it provides increased durability and abrasion resistance for diverse material compatibility.
  • Designed for blind hole tapping
  • Suitable for steel, stainless steel, aluminum, brass, bronze, copper, and cast iron
  • Features a 45° helix angle for effective chip evacuation
  • Coated with TiCN for enhanced performance
  • Includes an extended neck design
  • ANSI standard dimensions with H2 pitch diameter limits and 3B class of fit
  • Straight shank with square drive