Learn More
Grainger WIDIA Spiral Flute Tap, M8x1.25, Bottoming, High Speed Steel, 3 Flutes
Catalog No. 502771823
Encompass_Preferred
This M8x1.25 metric spiral flute tap is designed for efficient threading in non-ferrous metal applications. Manufactured from high-speed steel, this uncoated tool features a bottoming chamfer and is optimized for blind hole tapping requirements.
  • Constructed from durable high-speed steel for reliable performance
  • Configured with 3 flutes for effective chip evacuation
  • Features a D5 thread limit and 6H class of fit for precise tolerance control
  • Designed for right hand thread direction and rotation
  • Suitable for use in non-ferrous metal material applications
Catalog No. 50-277-1823 Supplier Grainger Supplier No. 13X326
This M8x1.25 metric spiral flute tap is designed for efficient threading in non-ferrous metal applications. Manufactured from high-speed steel, this uncoated tool features a bottoming chamfer and is optimized for blind hole tapping requirements.
  • Constructed from durable high-speed steel for reliable performance
  • Configured with 3 flutes for effective chip evacuation
  • Features a D5 thread limit and 6H class of fit for precise tolerance control
  • Designed for right hand thread direction and rotation
  • Suitable for use in non-ferrous metal material applications