Learn More
Grainger Greenfield Threading Spiral Point Tap, 1/4 in-20 UNC, H2, 2B, 2 Flute, 1 ea
Catalog No. 502753938
Encompass_Preferred
This spiral point tap is a precision-engineered tool designed for efficient thread cutting in through-hole applications. Constructed from high speed steel, it provides durability and consistent performance when working with materials including steel, aluminum, brass, bronze, and copper.
  • Designed for through-hole tapping applications
  • Spiral point geometry facilitates chip clearance for cleaner threading
  • Constructed from high speed steel for reliable tool life
  • Features a 1/4 in. shank diameter and 2-1/2 in. overall length
  • Plug chamfer style for versatile threading operations
  • Meets USCTI Table 302A standards
Catalog No. 50-275-3938 Supplier Grainger Supplier No. 15J837
This spiral point tap is a precision-engineered tool designed for efficient thread cutting in through-hole applications. Constructed from high speed steel, it provides durability and consistent performance when working with materials including steel, aluminum, brass, bronze, and copper.
  • Designed for through-hole tapping applications
  • Spiral point geometry facilitates chip clearance for cleaner threading
  • Constructed from high speed steel for reliable tool life
  • Features a 1/4 in. shank diameter and 2-1/2 in. overall length
  • Plug chamfer style for versatile threading operations
  • Meets USCTI Table 302A standards