Learn More
Grainger Chicago-Latrobe Spiral Point Tap, #10-24 UNC, 3 Flute, Plug Chamfer, 1 ea
Catalog No. 502756358
Encompass_Preferred
The spiral point tap is a precision cutting tool designed for threading through holes in steel and stainless steel applications. Engineered from high-speed steel with cobalt (HSS-E), this tap provides enhanced durability and abrasion resistance for demanding machining tasks. Its specialized 3-flute geometry facilitates efficient chip evacuation, while the plug chamfer ensures stability during the tapping process.
  • Features 3-flute design for improved chip flow
  • Constructed from cobalt-enriched HSS-E for increased abrasion resistance and tool life
  • Designed for through hole applications in steel and stainless steel (ISO codes P and M)
  • Precision ground to H3 pitch diameter limits with a 2B class of fit
  • Includes extended neck for versatility
Catalog No. 50-275-6358 Supplier Grainger Supplier No. 15J339
The spiral point tap is a precision cutting tool designed for threading through holes in steel and stainless steel applications. Engineered from high-speed steel with cobalt (HSS-E), this tap provides enhanced durability and abrasion resistance for demanding machining tasks. Its specialized 3-flute geometry facilitates efficient chip evacuation, while the plug chamfer ensures stability during the tapping process.
  • Features 3-flute design for improved chip flow
  • Constructed from cobalt-enriched HSS-E for increased abrasion resistance and tool life
  • Designed for through hole applications in steel and stainless steel (ISO codes P and M)
  • Precision ground to H3 pitch diameter limits with a 2B class of fit
  • Includes extended neck for versatility