Learn More
Grainger Greenfield Threading Spiral Point Tap, 3/4-10 UNC, H3, High Speed Steel, 1 ea
Catalog No. 502754796
Encompass_Preferred
This spiral point tap is a precision-engineered cutting tool designed for efficient and reliable thread creation in both blind and through-hole applications. Constructed from high-speed steel and featuring a bright, uncoated finish, it provides excellent wear resistance and performance across various materials including steel and non-ferrous metals.
  • Designed for 3/4-10 UNC thread creation
  • Features a plug chamfer style with 3 flutes
  • Engineered for right-hand cutting applications
  • Meets USCTI Table 302A standards
  • Equipped with an H3 pitch diameter limit
  • Features a straight shank with square end for secure machine mounting
Catalog No. 50-275-4796 Supplier Grainger Supplier No. 15J882
This spiral point tap is a precision-engineered cutting tool designed for efficient and reliable thread creation in both blind and through-hole applications. Constructed from high-speed steel and featuring a bright, uncoated finish, it provides excellent wear resistance and performance across various materials including steel and non-ferrous metals.
  • Designed for 3/4-10 UNC thread creation
  • Features a plug chamfer style with 3 flutes
  • Engineered for right-hand cutting applications
  • Meets USCTI Table 302A standards
  • Equipped with an H3 pitch diameter limit
  • Features a straight shank with square end for secure machine mounting