Learn More
Grainger Chicago-Latrobe Spiral Point Machine Tap, HSS, Tin, #5-40 UNC, 1 ea
Catalog No. 502754777
Encompass_Preferred
This spiral point tap is a high-speed steel (HSS) machine tap designed for thread cutting in steel and stainless steel applications. It features a titanium nitride (TiN) coating for improved wear resistance and extended tool life. This #5-40 UNC tap is built to USCTI Table 302A standards with a plug chamfer and two flutes for efficient chip management in through-hole operations.
  • High-speed steel construction ensures durability and performance
  • TiN coating reduces friction and increases surface hardness
  • Spiral point geometry provides superior chip control by pushing chips forward
  • Plug chamfer design allows for versatile use in through-hole threading
  • Straight shank with square end for secure machine holding
Catalog No. 50-275-4777 Supplier Grainger Supplier No. 15J891
This spiral point tap is a high-speed steel (HSS) machine tap designed for thread cutting in steel and stainless steel applications. It features a titanium nitride (TiN) coating for improved wear resistance and extended tool life. This #5-40 UNC tap is built to USCTI Table 302A standards with a plug chamfer and two flutes for efficient chip management in through-hole operations.
  • High-speed steel construction ensures durability and performance
  • TiN coating reduces friction and increases surface hardness
  • Spiral point geometry provides superior chip control by pushing chips forward
  • Plug chamfer design allows for versatile use in through-hole threading
  • Straight shank with square end for secure machine holding