Learn More
Grainger Greenfield Threading Spiral Point Tap, #6-40 UNF, H2, 2B, 2 Flute, 1 ea
Catalog No. 502753826
Encompass_Preferred
This spiral point tap is a high-performance machine tap constructed from high speed steel. Designed for efficiency, this tool features ground threads and is intended for right-hand cutting applications in diverse materials including steel and non-ferrous metals like aluminum.
  • Plug chamfer type for versatile threading tasks
  • Bright finish for reduced friction and improved chip flow
  • H2 thread limit and 2B class of fit for precision thread accuracy
  • Two-flute design with straight flutes and a square end feature for secure attachment
  • Suitable for a wide range of industrial threading operations
Catalog No. 50-275-3826 Supplier Grainger Supplier No. 15J818
This spiral point tap is a high-performance machine tap constructed from high speed steel. Designed for efficiency, this tool features ground threads and is intended for right-hand cutting applications in diverse materials including steel and non-ferrous metals like aluminum.
  • Plug chamfer type for versatile threading tasks
  • Bright finish for reduced friction and improved chip flow
  • H2 thread limit and 2B class of fit for precision thread accuracy
  • Two-flute design with straight flutes and a square end feature for secure attachment
  • Suitable for a wide range of industrial threading operations