Learn More
Grainger Greenfield Threading Spiral Point Tap, M2 x 0.4, High Speed Steel, 2 Flute, 1 ea
Catalog No. 502756642
Encompass_Preferred
This M2 x 0.4 metric machine tap is a high speed steel spiral point tap designed for effective thread cutting in a variety of materials, including steel and non-ferrous metals like aluminum. Featuring a bright surface finish and a plug chamfer, this tool is engineered to deliver reliable performance in general-purpose machining applications.
  • Manufactured from high speed steel for durability and heat resistance.
  • Spiral point design facilitates efficient chip clearance by pushing chips forward through the hole.
  • Includes two straight flutes for stable cutting action.
  • Designed with a 6H thread tolerance and a D3 pitch diameter limit to meet precise thread standards.
  • Features an extended neck for improved reach in deep-hole threading tasks.
Catalog No. 50-275-6642 Supplier Grainger Supplier No. 15J801
This M2 x 0.4 metric machine tap is a high speed steel spiral point tap designed for effective thread cutting in a variety of materials, including steel and non-ferrous metals like aluminum. Featuring a bright surface finish and a plug chamfer, this tool is engineered to deliver reliable performance in general-purpose machining applications.
  • Manufactured from high speed steel for durability and heat resistance.
  • Spiral point design facilitates efficient chip clearance by pushing chips forward through the hole.
  • Includes two straight flutes for stable cutting action.
  • Designed with a 6H thread tolerance and a D3 pitch diameter limit to meet precise thread standards.
  • Features an extended neck for improved reach in deep-hole threading tasks.