Learn More
Grainger Greenfield Threading Spiral Point Tap, M3.5x0.6, HSS, Plug, D4/6H
Catalog No. 502756643
Encompass_Preferred
This high-speed steel (HSS) machine tap is designed for efficient, general-purpose threading in a wide variety of materials, including steel, aluminum, brass, bronze, and copper. Featuring a spiral point design, this tool effectively pushes chips forward to prevent clogging, which facilitates cleaner threads and higher production speeds in through-hole applications.
  • Features a plug chamfer style with a D4 tap limit and 6H thread tolerance.
  • Constructed from durable high-speed steel with a bright, uncoated finish.
  • Configured with 2 flutes and a straight shank with a square drive end.
  • Measures M3.5x0.6 with an overall length of 50.8 mm.
  • Engineered in accordance with USCTI Table 302A standards.
Catalog No. 50-275-6643 Supplier Grainger Supplier No. 15J813
This high-speed steel (HSS) machine tap is designed for efficient, general-purpose threading in a wide variety of materials, including steel, aluminum, brass, bronze, and copper. Featuring a spiral point design, this tool effectively pushes chips forward to prevent clogging, which facilitates cleaner threads and higher production speeds in through-hole applications.
  • Features a plug chamfer style with a D4 tap limit and 6H thread tolerance.
  • Constructed from durable high-speed steel with a bright, uncoated finish.
  • Configured with 2 flutes and a straight shank with a square drive end.
  • Measures M3.5x0.6 with an overall length of 50.8 mm.
  • Engineered in accordance with USCTI Table 302A standards.