Learn More
Electron Microscopy Sciences Spiral Saw Blades 1/0
SDP
Catalog No. 50949090
Encompass_Preferred
Swiss made saw blades. Cuts in any direction. Used for cutting rubber, plastics, wax, plaster, and similar materials.
Diameter: 0.0220" (.56mm)
Catalog No. 50-949-090 Supplier Electron Microscopy Sciences Supplier No. 7201501
Swiss made saw blades. Cuts in any direction. Used for cutting rubber, plastics, wax, plaster, and similar materials.
Diameter: 0.0220" (.56mm)