Learn More
Electron Microscopy Sciences Spiral Saw Blades 1
SDP
Catalog No. 50949091
Encompass_Preferred
Swiss made saw blades. Cuts in any direction. Used for cutting rubber, plastics, wax, plaster, and similar materials.
Diameter: 0.0240" (.61mm)
Catalog No. 50-949-091 Supplier Electron Microscopy Sciences Supplier No. 7201510
Swiss made saw blades. Cuts in any direction. Used for cutting rubber, plastics, wax, plaster, and similar materials.
Diameter: 0.0240" (.61mm)