Learn More
Electron Microscopy Sciences Spiral Saw Blades 2
SDP
Catalog No. 50949094
Encompass_Preferred
Swiss made saw blades. Cuts in any direction. Used for cutting rubber, plastics, wax, plaster, and similar materials.
Diameter: 0.0276" (.70mm)
Catalog No. 50-949-094 Supplier Electron Microscopy Sciences Supplier No. 7201520
Swiss made saw blades. Cuts in any direction. Used for cutting rubber, plastics, wax, plaster, and similar materials.
Diameter: 0.0276" (.70mm)