Learn More
Research Products International Corp SpiralPestle Grinder Kit, 100 each pestle and 1.5ml micro-tube
SDP
Catalog No. 501537759
Encompass
Unique SpiralPestle Grinder features screw-like pestle to homogenize culture cells, small tissue samples, centrifugal pellets, precipitates and other soft solids. SpiralPestle Grinder actively rotates and conveniently draws 10-100 mg of sample suspended in 200-500ul of extraction solution downward to bottom of 1.5 ml conical micro-tubes.
Ideal pestle disperses soft plant or animal tissues quickly and effectively by macerating and mechanically dispersing samples between the wall of the pestle and micro-tube. Adding glass grinding beads can further enhance sample grinding and its recommended for efficient cell lysis of tissues and microorganisms. Suppied 100 each (pestle and 1.5 ml micro-tube) plus black rubber coupler for use with BioVortexer.
Unique spiral design
Homogenizes cells and tissues
Disperses pellets, precipitates, and soft solids
Autoclavable Polypropylene
Catalog No. 50-153-7759 Supplier Research Products International Corp Supplier No. 299017
Unique SpiralPestle Grinder features screw-like pestle to homogenize culture cells, small tissue samples, centrifugal pellets, precipitates and other soft solids. SpiralPestle Grinder actively rotates and conveniently draws 10-100 mg of sample suspended in 200-500ul of extraction solution downward to bottom of 1.5 ml conical micro-tubes.
Ideal pestle disperses soft plant or animal tissues quickly and effectively by macerating and mechanically dispersing samples between the wall of the pestle and micro-tube. Adding glass grinding beads can further enhance sample grinding and its recommended for efficient cell lysis of tissues and microorganisms. Suppied 100 each (pestle and 1.5 ml micro-tube) plus black rubber coupler for use with BioVortexer.
Unique spiral design
Homogenizes cells and tissues
Disperses pellets, precipitates, and soft solids
Autoclavable Polypropylene