Learn More
Apexbio Technology LLC Spiramycin 8025-81-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063798
Encompass_Preferred
Spiramycin is a macrolide antibiotic characterized by a 16-membered lactone ring structure It exerts its antimicrobial action primarily by binding to the bacterial 50S ribosomal subunit thereby inhibiting protein synthesis Spiramycin is routinely utilized in microbiological and pharmacological research to investigate bacterial resistance mechanisms and ribosomal function It is employed in cell culture assays and antimicrobial susceptibility testing for bacterial strains aiding researchers studying macrolide-resistance pathways and antibiotic-target interactions Experimental data indicate spiramycin inhibits the growth of sensitive bacteria with IC50 values generally ranging between 0 1 10 g/mL depending on bacterial strain and assay conditions
Catalog No. 50-000-63798 Supplier Apexbio Technology LLC Supplier No. B18395.1
Spiramycin is a macrolide antibiotic characterized by a 16-membered lactone ring structure It exerts its antimicrobial action primarily by binding to the bacterial 50S ribosomal subunit thereby inhibiting protein synthesis Spiramycin is routinely utilized in microbiological and pharmacological research to investigate bacterial resistance mechanisms and ribosomal function It is employed in cell culture assays and antimicrobial susceptibility testing for bacterial strains aiding researchers studying macrolide-resistance pathways and antibiotic-target interactions Experimental data indicate spiramycin inhibits the growth of sensitive bacteria with IC50 values generally ranging between 0 1 10 g/mL depending on bacterial strain and assay conditions