Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spiramycin, G-Biosciences
SCI_ED

Catalog No. S139811
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139811 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139811 Supplier G-Biosciences Supplier No. RC1103
Add to Cart
Add to Cart

Spiramycin is a broad-spectrum macrolide antibiotic effective against gram-negative and gram-positive bacteria as well as some Toxoplasma parasites.

Spiramycin is a mixture of spiramycin I, II, and III and has been used to study sepsis in mice.

Chemical Identifiers

CAS 8025-81-8

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 5 g
Percent Purity >95%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.