Learn More
Medchemexpress LLC Spiramycin I | 24916-50-5 | 1 ML
SDP
Catalog No. 5000417591
Encompass_Preferred
Spiramycin I is a macrolide antibiotic and antiparasitic, acting by binding to the 50S subunit of the ribosome to block peptide chain synthesis. It demonstrates inhibitory activity against various Gram-positive bacteria and exhibits cytotoxicity in several cancer cell lines. This product is intended for research use only.
  • Inhibits Gram-positive bacteria such as *B. subtilis*, *M. luteus*, *S. aureus*, *S. epidermidis*, and *S. pneumoniae*
  • Exhibits cytotoxicity in various cancer cell lines including HeLa, KB, MCF-7, HepG2, and U87
  • Intended for research use only
Catalog No. 50-004-17591 Supplier Medchemexpress LLC Supplier No. HYN714110MM/1ML
Spiramycin I is a macrolide antibiotic and antiparasitic, acting by binding to the 50S subunit of the ribosome to block peptide chain synthesis. It demonstrates inhibitory activity against various Gram-positive bacteria and exhibits cytotoxicity in several cancer cell lines. This product is intended for research use only.
  • Inhibits Gram-positive bacteria such as *B. subtilis*, *M. luteus*, *S. aureus*, *S. epidermidis*, and *S. pneumoniae*
  • Exhibits cytotoxicity in various cancer cell lines including HeLa, KB, MCF-7, HepG2, and U87
  • Intended for research use only