Learn More
Medchemexpress LLC Spiramycin I | 24916-50-5 | 10 MG
SDP
Catalog No. 5000417546
Encompass_Preferred
Spiramycin I is a macrolide antibiotic and antiparasitic that inhibits protein synthesis by binding to the 50S ribosomal subunit in bacteria. It is effective against various Gram-positive bacteria and protozoal infections, including Toxoplasma gondii. It can also prevent the placental transmission of toxoplasmosis.
  • Antibiotic and antiparasitic agent
  • Inhibits Gram-positive bacteria
  • Effective against protozoal infections
  • Prevents placental transmission of toxoplasmosis
Catalog No. 50-004-17546 Supplier Medchemexpress LLC Supplier No. HYN714110MG
Spiramycin I is a macrolide antibiotic and antiparasitic that inhibits protein synthesis by binding to the 50S ribosomal subunit in bacteria. It is effective against various Gram-positive bacteria and protozoal infections, including Toxoplasma gondii. It can also prevent the placental transmission of toxoplasmosis.
  • Antibiotic and antiparasitic agent
  • Inhibits Gram-positive bacteria
  • Effective against protozoal infections
  • Prevents placental transmission of toxoplasmosis