Learn More
Medchemexpress LLC Spiramycin I | 24916-50-5 | 100 MG
SDP
Catalog No. 5000417615
Encompass_Preferred
Spiramycin I is a macrolide antibiotic and antiparasitic. It binds to the 50S subunit of the ribosome, blocking the channel for peptide chain synthesis, which leads to inhibitory activity against Gram-positive bacteria. Spiramycin I inhibits B. subtilis, M. luteus, S. aureus, S. epidermidis, and S. pneumoniae with MICs ranging from 0.5-8 μg/mL. It also exhibits cytotoxicity in cancer cells (HeLa, KB, MCF-7, HepG2, and U87) with IC50 values between 30.51 and 34.41 μM.
  • Binds to the 50S subunit of the ribosome
  • Blocks peptide chain synthesis
  • Inhibitory activity against Gram-positive bacteria
  • Inhibits various bacteria including B. subtilis, M. luteus, S. aureus, S. epidermidis, and S. pneumoniae
  • Exhibits cytotoxicity in cancer cells
  • For research use only
Catalog No. 50-004-17615 Supplier Medchemexpress LLC Supplier No. HYN7141100MG
Spiramycin I is a macrolide antibiotic and antiparasitic. It binds to the 50S subunit of the ribosome, blocking the channel for peptide chain synthesis, which leads to inhibitory activity against Gram-positive bacteria. Spiramycin I inhibits B. subtilis, M. luteus, S. aureus, S. epidermidis, and S. pneumoniae with MICs ranging from 0.5-8 μg/mL. It also exhibits cytotoxicity in cancer cells (HeLa, KB, MCF-7, HepG2, and U87) with IC50 values between 30.51 and 34.41 μM.
  • Binds to the 50S subunit of the ribosome
  • Blocks peptide chain synthesis
  • Inhibitory activity against Gram-positive bacteria
  • Inhibits various bacteria including B. subtilis, M. luteus, S. aureus, S. epidermidis, and S. pneumoniae
  • Exhibits cytotoxicity in cancer cells
  • For research use only