Learn More
Medchemexpress LLC Spiramycin I | 24916-50-5 | 25 MG
SDP
Catalog No. 5000421521
Encompass_Preferred
Spiramycin I is a macrolide antibiotic and antiparasitic. It works by binding to the 50S subunit of the ribosome, which blocks the channel for peptide chain synthesis. This mechanism leads to inhibitory activity against Gram-positive bacteria. Additionally, Spiramycin I has shown cytotoxicity in various cancer cell lines.
  • Macrolide antibiotic and antiparasitic
  • Binds to 50S ribosomal subunit to block peptide synthesis
  • Exhibits inhibitory activity against Gram-positive bacteria
  • Demonstrates cytotoxicity in cancer cells
Catalog No. 50-004-21521 Supplier Medchemexpress LLC Supplier No. 25MGHYN7141
Spiramycin I is a macrolide antibiotic and antiparasitic. It works by binding to the 50S subunit of the ribosome, which blocks the channel for peptide chain synthesis. This mechanism leads to inhibitory activity against Gram-positive bacteria. Additionally, Spiramycin I has shown cytotoxicity in various cancer cell lines.
  • Macrolide antibiotic and antiparasitic
  • Binds to 50S ribosomal subunit to block peptide synthesis
  • Exhibits inhibitory activity against Gram-positive bacteria
  • Demonstrates cytotoxicity in cancer cells