Learn More
Medchemexpress LLC Spiramycin I | 24916-50-5 | 5 MG
SDP
Catalog No. 5000417691
Encompass_Preferred
Spiramycin I is a macrolide antibiotic and antiparasitic that functions by binding to the 50S subunit of the ribosome, thereby blocking peptide chain synthesis. It is intended for research use only and not for sale to patients.
  • Exhibits inhibitory activity against Gram-positive bacteria.
  • Inhibits B. subtilis, M. luteus, S. aureus, S. epidermidis, and S. pneumoniae.
  • Shows cytotoxicity in cancer cells HeLa, KB, MCF-7, HepG2, and U87.
Catalog No. 50-004-17691 Supplier Medchemexpress LLC Supplier No. HYN71415MG
Spiramycin I is a macrolide antibiotic and antiparasitic that functions by binding to the 50S subunit of the ribosome, thereby blocking peptide chain synthesis. It is intended for research use only and not for sale to patients.
  • Exhibits inhibitory activity against Gram-positive bacteria.
  • Inhibits B. subtilis, M. luteus, S. aureus, S. epidermidis, and S. pneumoniae.
  • Shows cytotoxicity in cancer cells HeLa, KB, MCF-7, HepG2, and U87.