Learn More
Medchemexpress LLC Spiramycin I | 24916-50-5 | 50 MG
SDP
Catalog No. 5000417202
Encompass_Preferred
Spiramycin I is a macrolide antibiotic and antiparasitic intended for research use only. It functions by binding to the 50S subunit of the ribosome, which blocks the synthesis of peptide chains.
  • Acts as a macrolide antibiotic and antiparasitic
  • Exhibits inhibitory activity against Gram-positive bacteria, including B. subtilis, M. luteus, S. aureus, S. epidermidis, and S. pneumoniae with MICs ranging from 0.5-8 μg/mL
  • Shows cytotoxicity in cancer cells such as HeLa, KB, MCF-7, HepG2, and U87, with IC50 values between 30.51 and 34.41 μM
Catalog No. 50-004-17202 Supplier Medchemexpress LLC Supplier No. HYN714150MG
Spiramycin I is a macrolide antibiotic and antiparasitic intended for research use only. It functions by binding to the 50S subunit of the ribosome, which blocks the synthesis of peptide chains.
  • Acts as a macrolide antibiotic and antiparasitic
  • Exhibits inhibitory activity against Gram-positive bacteria, including B. subtilis, M. luteus, S. aureus, S. epidermidis, and S. pneumoniae with MICs ranging from 0.5-8 μg/mL
  • Shows cytotoxicity in cancer cells such as HeLa, KB, MCF-7, HepG2, and U87, with IC50 values between 30.51 and 34.41 μM