Learn More
Selleck Chemical LLC Spiramycin I
SDP
Catalog No. 5000003717
Encompass_Preferred
Spiramycin I (Foromacidin A) is a main component of spiramycin which is a macrolide antimicrobial agent with broad spectrum antibiotic activity
Catalog No. 50-000-03717 Supplier Selleck Chemical LLC Supplier No. S57445MG
Spiramycin I (Foromacidin A) is a main component of spiramycin which is a macrolide antimicrobial agent with broad spectrum antibiotic activity