Learn More
Cayman Chemical Spirapril (hydrochloride), Purity: ≥98%, CAS: 94841-17-5, 50 mg
SDP
Catalog No. 502918582
Encompass_Preferred
An ACE inhibitor (IC50 = 67 nM) and a prodrug form of spiraprilat; inhibits the angiotensin I-induced pressor response in rats and dogs (ID50s = 16 and 262 µg/kg, respectively); decreases systolic blood pressure and left ventricular hypertrophy and induces angiogenesis in spontaneously hypertensive rats 5 mg/kg
Catalog No. 50-291-8582 Supplier Cayman Chemical Supplier No. 3927850 MG
An ACE inhibitor (IC50 = 67 nM) and a prodrug form of spiraprilat; inhibits the angiotensin I-induced pressor response in rats and dogs (ID50s = 16 and 262 µg/kg, respectively); decreases systolic blood pressure and left ventricular hypertrophy and induces angiogenesis in spontaneously hypertensive rats 5 mg/kg